VWR® Proline, Manual Dispensers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-2096EA 186 DKK
115-2096 115-2097
VWR® Proline, Manual Dispensers
Sæbedispensere og Lotion Sæbedispensere
These VWR® manual dispensers are durable, wall-mountable and for use with Deb Stoko® soaps and cleansers. Ideal for use in offices, medical rooms, laboratories, food services and catering.
  • Large push-button for one-handed or elbow operation
  • Delivers the exact amount required, controlling usage and reducing waste
  • Enables quick and easy cartridge replacement within seconds
  • Reduces maintenance time and costs
  • Allows no ingress of air into the cartridge during use
  • Helps prevent product contamination
Suitable for use with Deb Stoko® Refresh™ FOAM 1 litre cartridges.