Test kits, photometric, iron, LCK cuvette tests
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LCK321 LCK521 LCK320
705-3043EA 1070 DKK
705-3043 705-3044 705-3045
Test kits, photometric, iron, LCK cuvette tests
Miljø test kit
Cuvette tests for water testing applications.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Shelf life (LCK321): 30 months from production date
Shelf life (LCK320 and LCK521): 12 months from production date
Storage conditions: 2 to 8 °C (keep refrigerated)
Certificeringer: LCK321 and LCK521: ISO 6332-1988, DIN 38406 E1-1
Information om tilbehør: Cuvette tests are officially approved for many legally required consent limits, with the help of Addista standard solutions and round-robin test solutions, they provide the assurance you need.