Transport Container with Attached Lid
Leverandør: MULTIROIR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DEMB32L/R DEMB86L/R440 DEMB86L/R340
216-1961EA 193 DKK
216-1961 216-1971 216-1970
Transport Container with Attached Lid
Bokse Opbevaringsbokse
Transport container, PP, with attached lid.
  • Rigid, robust and practical
  • Containers nest for maximum space usage with lids open
  • Available in different sizes
  • Stackable containers
  • Colour: Blue
  • Easy to clean interiors
These containers with robust lids are stackable when they are closed. The recesses of the hinged lid ensure high stability. With open lids, the containers can be stacked into one another to save space.