Moisturising hand and body lotion, Stokolan® Intense
Leverandør: Deb-STOKO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SIN100ML SIN1L RES1LDS-UK
115-2364EA 528 DKK
115-2364 115-2365 EVONRES1LDS-UK
Moisturising hand and body lotion, Stokolan® Intense
Hudpleje Cremer Hand and Body Lotions
Stokolan® Intense is a nurturing, regenerative and barrier active creamy lotion containing grapeseed oil, ceramid complex, allantoin, ginkgo biloba, urea and beeswax. The lotion contains a high amount of lipids, 99% of natural origin, reducing water loss and enhancing skin elasticity. Suitable for facial and body care.
  • Ideal for very dry and barrier-damaged skin, as well as for the prevention of attrition dermatitis
  • Contains 10% grapeseed oil: Regulates the skin's own lipid and moisture balance
  • Contains urea und sorbitol: Excellent skin hydrating and moisturising properties
  • Dermatologically confirmed skin compatibility
  • Silicone-free and paint compatible
Certificeringer: ECARF Quality Seal for allergy friendliness
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904