Nitrile gloves, Microflex® 93-243
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
93-243/8.5-9 93-243/9.5-10 93-243/10.5-11 93-243/6.5-7 93-243/7.5-8 93-243/5.5-6
112-4579EA 891 DKK
112-4579 112-4580 112-4575 112-4577 112-4578 112-4576
Nitrile gloves, Microflex® 93-243
Handsker
These disposable gloves are made with a soft nitrile formulation. This formulation offers greater puncture resistance and chemical resistance than natural rubber latex.
  • Designed with elbow-length cuff (400 mm) to provide additional protection to the upper arm and extra security against cross-contamination
  • Improved cuff design helps prevent gloves from rolling down
  • Textured fingers and palm ensure strong grip in a variety of applications
  • Gloves are packed in polybags to help protect product integrity
Ideal for use in the chemical and automotiveMore Product Information