Homogeniser, WiseTis® HG-15A
Leverandør: witeg Labortechnik
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
DH.WHG02016 DH.WHG02017
WITGDH.WHG02016EA 8410 DKK
WITGDH.WHG02016 WITGDH.WHG02017
Homogeniser, WiseTis® HG-15A
Homogenisatorer Homogenisatorer, bordmodeller
Homogenisers with direct controllers for liquid and tissue samplers.
  • High efficiency dispersing tool made of stainless steel and PTFE
  • Over-heat protection
  • High speed and low noise: max. 27000 min⁻¹ (without load)
These homogenisers has analog control system with body made of aluminum die-casting and ABS.