Compression fittings
Leverandør: HAMILTON BONADUZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
55752-01 55753-01 55750-01 55751-01
HAMI55752-01EA 651 DKK
HAMI55752-01 HAMI55753-01 HAMI55750-01 HAMI55751-01
Compression fittings
Fittings og stik
The compression fittings and adaptors are designed to connect Hamilton removable needle (RN) and knurled hub (KH) syringes to glass capillary tubing and PEEK tubing. The connectors can be combined in a variety of different configurations to meet most connection needs.
Applications include: Neuroscience injections, patch clamp, cell transplantation, stem cell research, corrosive chemical dispensing, infusion pump injections, and mass spectrometry infusion pumps.