Crucibles, low form, quartz
Leverandør: TECHNICAL QUARTZ SOLUTIONS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
75052 75056 75058 75060 75062 75064 75066 75072
TSLA75052EA 181 DKK
TSLA75052 TSLA75056 TSLA75058 TSLA75060 TSLA75062 TSLA75064 TSLA75066 TSLA75072
Crucibles, low form, quartz
Digler
Quartz, transparent, glazed.
  • Lids for low form crucible, ring handle
Bestillingsoplysninger: Crucible and lid have to be ordered separately.