Microplates, micro-volume, SpectraDrop™
Leverandør: Molecular Devices
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
0200-6262 0200-6267 0200-6263 0200-6264 0200-6265 0200-6266
MLDV0200-6262EA 0 DKK
MLDV0200-6262 MLDV0200-6267 MLDV0200-6263 MLDV0200-6264 MLDV0200-6265 MLDV0200-6266
Microplates, micro-volume, SpectraDrop™
Mikroplader HTS mikroplader
SpectraDrop™ micro-volume microplate consists of aspecially designed adapter, PTFE-coated bottom slide, and top slide with evaporation-reducing spacers.
  • Handle volumes down to 2 µl
  • Used to quantify DNA, RNA and protein
  • Cleaning convenience with hasslefree handling
  • PTFE-coated to ensure precise droplet placement
  • Slide design is compatible with 8- and/or 16-channel pipettors
These microplates comes in two density configurations, 24-wells per plate and 64-wells per plate, with two volume options, 2 or 4 µl, allowing users to select the configurationMore Product Information