Anti-CD3 Rabbit Polyclonal Antibody

Leverandør: IMMUNOLOGIC
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ILP3333-C01 ILP3333-C05 ILP3333-C1 S/ILP3333-R2
ILP3333-C01EA 1670 DKK
ILP3333-C01 ILP3333-C05 VWRKILP3333-C1 VWRKS/ILP3333-R2
Anti-CD3 Rabbit Polyclonal Antibody
Antistoffer
Anti-CD3 antibody has been considered the best all around T-cell marker. This antibody reacts with an antigen present in early thymocytes. The positive staining of this marker may represent a sign of early commitment to the T-cell lineage.

  • Visualization: membranous
  • Control: tonsil
  • Dilution range: 1:100-1:1000

Pretreatment: Heat induced epitope retrieval in 10 mM citrate buffer , pH 6,0 or in 50 mM Tris buffer pH 9,5 for 20 minutes is required for IHC staining on formalin-fixed, paraffin embedded tissue sections.

Type: Primary
Antigen: CD3
Clonality:More Product Information
Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR