Stirring Rods, Thermo Fisher Scientific
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42294.NK 42293.NK 42292.NT 42293.NT 42294.NT
42294.NKEA 350 DKK
42294.NK 42293.NK 42292.NT 42293.NT 42294.NT
Stirring Rods, Thermo Fisher Scientific
Omrørere Omrørestave
The fused quartz rods are manufactured by fusing naturally occurring crystalline silica.
  • Fused quartz has high chemical purity, high corrosion resistance and high melting point
  • Quartz is extremely hard, with a low coefficient of thermal expansion, excellent electrical insulation qualities and optical transmission from ultra-violet to infra-red