2-Brom-4-thiazolcarboxylsyre 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
2-Bromo-1,3-thiazole-4-carboxylic acid, 2-Bromothiazole-4-carboxylic acid
454600010
454600050
ACRO454600010EA
1020
DKK
ACRO454600010
ACRO454600050
2-Brom-4-thiazolcarboxylsyre 98%
2-Brom-4-thiazolcarboxylsyre
Formel:
C₄H₂BrNO₂S Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04115729 CAS-nummer: 5198-88-9 |
Specifikationer Testresultater
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...