4-Methyl-1,3-thiazole-5-sulphonyl chloride 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 4-Methylthiazole-5-sulphonyl chloride

453640010 453640050
ACRO453640010EA 1400 DKK
ACRO453640010 ACRO453640050
4-Methyl-1,3-thiazole-5-sulphonyl chloride 97%
4-Methyl-1,3-thiazole-5-sulphonyl chloride
Formel: C₄H₄ClNO₂S₂
Smeltepunkt: 34…36 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09864968
CAS-nummer: 953070-51-4
UN nummer: 3261
ADR: 8,III

Bestil nu

Specifikationer Testresultater

Appearance White to yellow to brown Low melting solid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR