4-Methoxy-N-methylbenzylamine 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
451730010
451730100
ACRO451730010EA
458
DKK
ACRO451730010
ACRO451730100
4-Methoxy-N-methylbenzylamine 95%
4-Methoxy-N-methylbenzylamine
Formel:
C₉H₁₃NO Kogepunkt: 222 °C Massefylde: 1,008 g/cm³ Flammepunkt: 109 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD04115407 CAS-nummer: 702-24-9 ADR: 8,III |
Specifikationer Testresultater
Appearance | Colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5290 to 1.5310 (20°C, 589 nm) |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...