L-Lysine acetate salt 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: L-LYSINE, MONOACETATE, (2S)-2,6-diaminohexanoic acid, L-Lysine acetate, L-Lysine Acetate Salt

451630250 451631000 451635000
ACRO451630250EA 349 DKK
ACRO451630250 ACRO451631000 ACRO451635000
L-Lysine acetate salt 97%
L-Lysine acetate salt
Formel: C₈H₁₈N₂O₄
Molekylvægt: 206.242 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00039069
CAS-nummer: 57282-49-2

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Assay ≥96.0 %
Specific optical rotation +8° to +10° (20°C, 589 nm) (c=10, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR