Kaliumhydrogenkarbonat, extra pure

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Potassium bicarbonate

450640010 450640050 450640250
ACRO450640010EA 396 DKK
ACRO450640010 ACRO450640050 ACRO450640250
Kaliumhydrogenkarbonat, extra pure
Kaliumhydrogenkarbonat
Formel: KHCO₃
Molekylvægt: 100.12 g/mol
Smeltepunkt: 292 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011402
CAS-nummer: 298-14-6
Merck Index: 13,07691

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Solid
Titration with HCl ≥98.5 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR