Methyl2-methylnicotinat 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
methyl 2-methylpyridine-3-carboxylate, Methyl 2-methylnicotinate
449250050
449250250
ACRO449250050EA
746
DKK
ACRO449250050
ACRO449250250
Methyl2-methylnicotinat 97%
Methyl2-methylnicotinat
Formel:
C₈H₉NO₂ Molekylvægt: 151.16 g/mol Kogepunkt: 102…104 °C (10 mmHg) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00051961 CAS-nummer: 65719-09-7 |
Specifikationer Testresultater
Appearance | Colorless to yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5145 to 1.5185 (20°C, 589 nm) |
Water | ≤1.5 % |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...