Cortisone acetate 98+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Cortisone 21-acetate

448960050 448960250
ACRO448960050EA 991 DKK
ACRO448960050 ACRO448960250
Cortisone acetate 98+%
Cortisone acetate
Formel: C₂₃H₃₀O₆
Molekylvægt: 402.49 g/mol
Smeltepunkt: 237…240 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003609
CAS-nummer: 50-04-4

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥98.0 %
Specific optical rotation +207° to +220° (20°C, 589 nm) (c=1, dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR