Prednison 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

448950050 448950250
ACRO448950050EA 1560 DKK
ACRO448950050 ACRO448950250
Prednison 98%
Prednison
Formel: C₂₁H₂₆O₅
Smeltepunkt: 225…229 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003608
CAS-nummer: 53-03-2

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥97.5 %
Specific optical rotation +167° to +175° (20°C, 589 nm) (c= 0.5, dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR