Molybdentrioxid 99.5+% ACS
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Molybdenum trioxide
447401000
447405000
ACRO447401000EA
459
DKK
ACRO447401000
ACRO447405000
Molybdentrioxid 99.5+% ACS
Molybdentrioxid
Formel:
MoO₃ Kogepunkt: 1155 °C (760 mmHg) Smeltepunkt: 795 °C Massefylde: 4,69 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003469 CAS-nummer: 1313-27-5 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | White to yellow to green to grey Powder |
Titration | ≥99.5 % (Precipitation titration of molybdate) |
Chloride (Cl) | ≤0.002 % |
Nitrate (NO3) | Passes test (Limit about 0.003%) |
Phosphate (PO4) | ≤5 ppm |
Sulfate (SO4) | ≤0.02 % |
AsO4, PO4, SiO4 (as SiO2) | ≤0.001 % |
Ammonium (NH4) | ≤0.002 % |
Heavy metals (as Pb) | ≤0.005 % |
Insoluble matter | ≤0.01 % (In dilute NH4OH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...