5-(Chlormethyl)uracil 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
5-(Chloromethyl)pyrimidine-2,4(1H,3H)-dione
446190010
446190050
ACRO446190010EA
1660
DKK
ACRO446190010
ACRO446190050
5-(Chlormethyl)uracil 97%
5-(Chlormethyl)uracil
Formel:
C₅H₅ClN₂O₂ Smeltepunkt: >350 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00218445 CAS-nummer: 3590-48-5 |
Specifikationer Testresultater
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...