5-Chlor-3-pyridinboronsyre 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
445360010
445360050
ACRO445360010EA
2820
DKK
ACRO445360010
ACRO445360050
5-Chlor-3-pyridinboronsyre 95%
5-Chlor-3-pyridinboronsyre
Formel:
C₅H₅BClNO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06798243 CAS-nummer: 872041-85-5 |
Specifikationer Testresultater
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
HPLC | ≥94.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...