2,4-Dimethoxy-5-pyrimidineboronic acid pinacol ester 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

444930010 444930050
ACRO444930010EA 945 DKK
ACRO444930010 ACRO444930050
2,4-Dimethoxy-5-pyrimidineboronic acid pinacol ester 97%
2,4-Dimethoxy-5-pyrimidineboronic acid pinacol ester
Formel: C₁₂H₁₉BN₂O₄
Smeltepunkt: 104…109 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06795654
CAS-nummer: 936250-17-8

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Solid
Infrared spectrum Conforms
Melting point 104 to 109 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR