FMOC-O-tert-butyl-L-tyrosine 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-α-Fmoc-o-t-butyl-L-Tyrosine, N-Fmoc-O-tert-butyl-L-tyrosine, Nalpha-Fmoc-O-tert-butyl-L-tyrosine, O-tert-Butyl-Nalpha-Fmoc-L-tyrosine, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine, Fmoc-Tyr(tBu)-OH

439090250 439090100
ACRO439090250EA 574 DKK
ACRO439090250 ACRO439090100
FMOC-O-tert-butyl-L-tyrosine 98%
FMOC-O-tert-butyl-L-tyrosine
Formel: C₂₈H₂₉NO₅
Smeltepunkt: 150…156 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00037129
CAS-nummer: 71989-38-3

Bestil nu

Specifikationer Testresultater

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -27° to -31° (20°C, 589 nm) (c=1 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR