Cyclopropanesulphonyl chloride 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

439020010 439020050
ACRO439020010EA 2630 DKK
ACRO439020010 ACRO439020050
Cyclopropanesulphonyl chloride 97%
Cyclopropanesulphonyl chloride
Formel: C₃H₅ClO₂S
Kogepunkt: 60 °C (2Torr)
Massefylde: 1,38 g/cm³
Flammepunkt: 96 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01631933
CAS-nummer: 139631-62-2
UN nummer: 3265
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR