(3S)-(+)-1-Benzyl-3-aminopyrrolidine 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (S)-1-Benzyl-3-aminopyrrolidine, (S)-(+)-1-Benzyl-3-aminopyrrolidine

437880010 437880050
ACRO437880010EA 786 DKK
ACRO437880010 ACRO437880050
(3S)-(+)-1-Benzyl-3-aminopyrrolidine 99%
(3S)-(+)-1-Benzyl-3-aminopyrrolidine
Formel: C₁₁H₁₆N₂
Molekylvægt: 176.26 g/mol
Kogepunkt: 108…115 °C (5 mmHg)
Massefylde: 1.02 g/cm³
Flammepunkt: 140 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00082766
CAS-nummer: 114715-38-7
UN nummer: 2735
ADR: 8 III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥98.5 %
Specific optical rotation +1° to +3° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR