Sodium hexanoate 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
437540100
437540250
ACRO437540100EA
708
DKK
ACRO437540100
ACRO437540250
Sodium hexanoate 99%
Sodium hexanoate
Formel:
C₆H₁₁NaO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00059056 CAS-nummer: 10051-44-2 |
Specifikationer Testresultater
Appearance | White Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...