Mangan(II)oxid 99.99% (trace metals basis)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Manganese(ii)oxide, Manganese oxide
436970010
436970100
ACRO436970010EA
315
DKK
ACRO436970010
ACRO436970100
Mangan(II)oxid 99.99% (trace metals basis)
Manganese(II) oxide
Formel:
MnO Smeltepunkt: 1650 °C Massefylde: 5,43 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00016226 CAS-nummer: 1344-43-0 |
Specifikationer Testresultater
Appearance | Green Crystalline powder and/or chunks |
Titration Complexometric | 76.7 to 78.2 % (Mn) |
Total trace metal impurities | ≤100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...