6-fluordipicolinsyre 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
436840010
436840050
ACRO436840010EA
1770
DKK
ACRO436840010
ACRO436840050
6-fluordipicolinsyre 97%
6-fluordipicolinsyre
Formel:
C₆H₄FNO₂ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD02181193 CAS-nummer: 402-69-7 |
Specifikationer Testresultater
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 135 to 142 °C |
Water | ≤1.0 % |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...