Naproxen sodium 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
436710050
436710250
ACRO436710050EA
737
DKK
ACRO436710050
ACRO436710250
Naproxen sodium 98%
Naproxen sodium
Formel:
C₁₄H₁₃NaO₃ Smeltepunkt: 259…262 °C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00058507 CAS-nummer: 26159-34-2 |
Specifikationer Testresultater
Appearance | Colorless to light yellow Powder |
Infrared spectrum | Conforms |
Specific optical rotation | -14.7° to -17.0° (20°C, 589 nm) (c=5, 1N NaOH) |
Titration with HClO4 | 98.0 to 102.0 % |
Free acid | ≤1.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...