Methyl-2-(hydroxymethyl)acrylate 85% stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 2-(hydroxymethyl)acrylate

Stabilization: 0.1 wt.% Stabilised with 4-methoxyphenol (MEHQ)

435900010 435900050
ACRO435900010EA 484 DKK
ACRO435900010 ACRO435900050
Methyl-2-(hydroxymethyl)acrylate 85% stabilised
Methyl-2-(hydroxymethyl)acrylate
Formel: C₅H₈O₃
Molekylvægt: 116.1152 g/mol
Kogepunkt: 71…72 °C (2Torr)
Massefylde: 1.129 g/cm³
Flammepunkt: 90 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD12545894
CAS-nummer: 15484-46-5

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
GC ≥84.0 %
NMR consistent with structure
Specific optical rotation +8° to +11° (20°C, 589 nm) (c=1,CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR