N-Carbobenzoxy-β-alanine 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

434810050 434810250
ACRO434810050EA 463 DKK
ACRO434810050 ACRO434810250
N-Carbobenzoxy-β-alanine 98%
N-Carbobenzoxy-beta-alanine
Formel: C₁₁H₁₃NO₄
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00037292
CAS-nummer: 2304-94-1

Bestil nu

Specifikationer Testresultater

Appearance White Powder
Infrared spectrum Conforms
Melting point 100 to 110 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR