4-Brom-2-chlorpyridin 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
434800050
434800250
ACRO434800050EA
784
DKK
ACRO434800050
ACRO434800250
4-Brom-2-chlorpyridin 97%
4-Bromo-2-chloropyridine
Formel:
C₅H₃BrClN Massefylde: 1,734 g/cm³ Flammepunkt: 104 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD03840756 CAS-nummer: 73583-37-6 |
Specifikationer Testresultater
Appearance | Clear colorless to white to yellow to orange Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...