N-Lauroylsarcosin natrium salt 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate
434370010
434370250
434371000
ACRO434370010EA
3370
DKK
ACRO434370010
ACRO434370250
ACRO434371000
N-Lauroylsarcosin natrium salt 95%
N-Lauroylsarcosin natrium salt
Formel:
H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa Molekylvægt: 293.38 g/mol Smeltepunkt: 45…47 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00042728 CAS-nummer: 137-16-6 UN nummer: 2811 ADR: 6.1 II Merck Index: 12,04379 |
|
Specifikationer Testresultater
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Water | ≤7 % |
Solubility | (10 % in water) Almost clear colorless to light yellow |
pH | 7.0 to 9.0 |
Titration with HClO4 | 94 to 106 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...