5-Amino-2-chloropyrimidine 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
434320010
434320050
ACRO434320010EA
1290
DKK
ACRO434320010
ACRO434320050
5-Amino-2-chloropyrimidine 95%
5-Amino-2-chloropyrimidine
Formel:
C₄H₄ClN₃ Smeltepunkt: 160 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD06412560 CAS-nummer: 56621-90-0 |
Specifikationer Testresultater
Appearance | Yellow to brown Crystalline powder or solid |
Infrared spectrum | Conforms |
GC | ≥94.0 % |
Specific optical rotation | +27° to +29° (20°C, 589 nm) (C=4, 5 N HCl) |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...