Ytterbium(III)trifluormethansulphonat, anhydrous 99.9% (trace metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Ytterbium (III) triflate

434050010 434050050
ACRO434050010EA 387 DKK
ACRO434050010 ACRO434050050
Ytterbium(III)trifluormethansulphonat, anhydrous 99.9% (trace metals basis)
Ytterbium(III)trifluormethansulphonat
Formel: C₃F₉O₉S₃Yb
Molekylvægt: 620.25 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD06200261
CAS-nummer: 54761-04-5
UN nummer: 3261
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Complexometric 26.8 to 29.0 % (Yb)
Total conc. of imp.as REO ≤1000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR