Ethylbenzen, anhydrous 99.8%, AcroSeal®
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Phenylethane
433800010
433801000
433800010.EA
1560
DKK
433800010.
433801000.
Ethylbenzen, anhydrous 99.8%, AcroSeal®
Ethylbenzen
Formel:
C₈H₁₀ Molekylvægt: 106.17 g/mol Kogepunkt: 136 °C (760 mmHg) Smeltepunkt: –95 °C Massefylde: 0.86 g/cm³ Flammepunkt: 22 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011647 CAS-nummer: 100-41-4 UN nummer: 1175 ADR: 3 II Merck Index: 13,03801 |
|
Specifikationer Testresultater
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Water | ≤0.005 % |
GC | ≥99.75 % |
Residue after evaporation | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...