2-Methoxybenzonitril 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

433160050 433160250
ACRO433160050EA 347 DKK
ACRO433160050 ACRO433160250
2-Methoxybenzonitril 99%
2-Methoxybenzonitril
Formel: C₈H₇NO
Kogepunkt: 256 °C (760 mmHg)
Smeltepunkt: 21…22 °C
Massefylde: 1,095 g/cm³ (15 °C)
Flammepunkt: 113 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00001783
CAS-nummer: 6609-56-9
UN nummer: 3276
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to light yellow Liquid or low melting solid
Infrared spectrum Conforms
GC ≥98.0 %
Residue after evaporation ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR