Natriumdihydrogenorthoperiodat 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Trisodium dihydrogenorthoperiodate, Sodium dihydroperiodate

432851000 432855000
ACRO432851000EA 617 DKK
ACRO432851000 ACRO432855000
Natriumdihydrogenorthoperiodat 98%
Sodium (para)periodate
Formel: H₂INa₃O₆
Smeltepunkt: >300 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00036146
CAS-nummer: 13940-38-0
UN nummer: 1479
ADR: 5.1,II

Bestil nu

Specifikationer Testresultater

Appearance White Powder
Titration Iodometric ≥97.5 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR