Methyltrimethoxysilan 97%, AcroSeal®
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
432640010
432641000
432640010.EA
795
DKK
432640010.
432641000.
Methyltrimethoxysilan 97%, AcroSeal®
Methyltrimethoxysilane
Formel:
C₄H₁₂O₃Si Kogepunkt: 102 °C (760 mmHg) Smeltepunkt: –50 °C Massefylde: 0,95 g/cm³ Flammepunkt: 11 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008342 CAS-nummer: 1185-55-3 UN nummer: 1993 ADR: 3,II |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3695 to 1.3715 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...