Thiopicolinamide 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Pyridine-2-carbothioamide, 2-Pyridinethioamide, 2-Pyridylthiocarboxamide

432470050 432470250
ACRO432470050EA 283 DKK
ACRO432470050 ACRO432470250
Thiopicolinamide 97%
Thiopicolinamide
Formel: C₆H₆N₂S
Molekylvægt: 138.19 g/mol
Smeltepunkt: 137…139 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00087576
CAS-nummer: 5346-38-3

Bestil nu

Specifikationer Testresultater

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 134 to 140 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR