Dichlorodiisopropylsilane 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
432360050
432360250
ACRO432360050EA
499
DKK
ACRO432360050
ACRO432360250
Dichlorodiisopropylsilane 97%
Dichlorodiisopropylsilane
Formel:
C₆H₁₄Cl₂Si Kogepunkt: 168…170 °C (760 mmHg) Massefylde: 1,026 g/cm³ Flammepunkt: 43 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00054895 CAS-nummer: 7751-38-4 UN nummer: 2986 ADR: 8,II |
Specifikationer Testresultater
Appearance | Colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4430 to 1.4450 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...