Thiophenol 99%, AcroSeal®
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Mercaptobenzene, Phenyl mercaptan, Benzenethiol
430721000
430728000
430721000.EA
285
DKK
430721000.
430728000.
Thiophenol 99%, AcroSeal®
Thiophenol
Formel:
C₆H₆S Molekylvægt: 110.18 g/mol Kogepunkt: 167…169 °C (760 mmHg) Smeltepunkt: –15 °C Massefylde: 1.078 g/cm³ Flammepunkt: 50 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004826 CAS-nummer: 108-98-5 UN nummer: 2337 ADR: 6.1 I Merck Index: 12,09492 |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5880 to 1.5900 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...