Benzofurazan-5-boronic acid pinacol ester 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]oxadiazole

430470010 430470050
ACRO430470010EA 864 DKK
ACRO430470010 ACRO430470050
Benzofurazan-5-boronic acid pinacol ester 97%
Benzofurazan-5-boronic acid pinacol ester
Formel: C₁₂H₁₅BN₂O₃
Molekylvægt: 246.07 g/mol
Smeltepunkt: 88…90 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09842717
CAS-nummer: 1073355-14-2

Bestil nu

Specifikationer Testresultater

Appearance White to yellow to tan Powder
Infrared spectrum Conforms
Melting point 86 to 91 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR