6-Bromisoquinolin 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

429460010 429460050
ACRO429460010EA 652 DKK
ACRO429460010 ACRO429460050
6-Bromisoquinolin 97%
6-Bromisoquinolin
Formel: C₉H₆BrN
MDL Number: MFCD04973299
CAS-nummer: 34784-05-9

Bestil nu

Specifikationer Testresultater

Appearance Orange to light brown Low melting solid
Infrared spectrum Conforms
Melting point 40 to 45 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR