Chloromethyl phenyl sulphone 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
((Chloromethyl)sulphonyl)benzene, 1-(Chloromethylsulphonyl)benzene, Phenyl Chloromethyl Sulphone
428740050
428740250
ACRO428740050EA
881
DKK
ACRO428740050
ACRO428740250
Chloromethyl phenyl sulphone 99%
Chloromethyl phenyl sulphone
Formel:
C₇H₇ClO₂S Smeltepunkt: 51…53 °C Flammepunkt: 113 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00007551 CAS-nummer: 7205-98-3 |
Specifikationer Testresultater
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 50 to 54 °C |
GC | ≥98.5 % |
NMR (1H-NMR) | Consistent with structure |
Free alkali (KOH) | ≤1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...