9-Bromo-9-phenylfluorene 97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
428610010
428610050
ACRO428610010EA
373
DKK
ACRO428610010
ACRO428610050
9-Bromo-9-phenylfluorene 97%
9-Bromo-9-phenylfluorene
Formel:
C₁₉H₁₃Br Smeltepunkt: 99…101 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00075522 CAS-nummer: 55135-66-5 UN nummer: 3261 ADR: 8,II |
Specifikationer Testresultater
Appearance | Yellow Powder |
Infrared spectrum | Conforms |
Melting point | 99 to 103 °C |
HPLC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...