4-Vinylphenyl acetate 96% stabilised
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Acetic Acid 4-Vinylphenyl Ester, 4-Acetoxystyrene
Stabilization:
200-300 ppm Stabilised with 4-methoxyphenol (MEHQ)
427000050
427000250
ACRO427000050EA
339
DKK
ACRO427000050
ACRO427000250
4-Vinylphenyl acetate 96% stabilised
4-Vinylphenyl acetate
Formel:
C₁₀H₁₀O₂ Molekylvægt: 162.19 g/mol Kogepunkt: 260 °C (760 mmHg) Smeltepunkt: 7…8 °C Massefylde: 1.06 g/cm³ Flammepunkt: 88 °C Opbevaringstemperatur: Fryser |
MDL Number:
MFCD00075734 CAS-nummer: 2628-16-2 |
Specifikationer Testresultater
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5375 to 1.5395 (20°C, 589 nm) |
GC | ≥95.0 % |
Stabilizer | 200 to 300 ppm MEHQ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...