Isoindoline 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

426280010 426280050
ACRO426280010EA 753 DKK
ACRO426280010 ACRO426280050
Isoindoline 97%
Isoindoline
Formel: C₈H₉N
Kogepunkt: 221 °C (760 mmHg)
Smeltepunkt: 17 °C
Massefylde: 1,05 g/cm³
Flammepunkt: 94 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00605324
CAS-nummer: 496-12-8

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless to yellow to pink to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5670 to 1.5710 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR