8-Anilino-1-naphthalensulfonsyre 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
8-Anilinonaphthalene-1-sulphonic acid, 8-(Phenylamino)naphthalene-1-sulphonic acid
401210050
401210250
401211000
ACRO401210050EA
478
DKK
ACRO401210050
ACRO401210250
ACRO401211000
8-Anilino-1-naphthalensulfonsyre 98%
8-Anilino-1-naphthalensulfonsyre
Formel:
C₁₆H₁₃NO₃S Molekylvægt: 299.35 g/mol Smeltepunkt: 183…183.5 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00003998 CAS-nummer: 82-76-8 |
Specifikationer Testresultater
Appearance | Dark green to brown to black Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...